Ethyl 3-bromobutyrate 95%

Supplier: Apollo Scientific

OR928737-1G OR928737-5G
APOSOR928737-1GEA 40.6 EUR
APOSOR928737-1G APOSOR928737-5G
Ethyl 3-bromobutyrate 95%
Ethyl 3-bromobutyrate
Formula: C₆H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00039175
CAS Number: 7425-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR