1-Benzylpyrazole-3-carbonitrile 95%

Supplier: Apollo Scientific

OR929440-1G OR929440-250MG
APOSOR929440-1GEA 632 EUR
APOSOR929440-1G APOSOR929440-250MG
1-Benzylpyrazole-3-carbonitrile 95%
1-Benzylpyrazole-3-carbonitrile
Formula: C₁₁H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD23381125
CAS Number: 1427475-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR