Ethyl 2,2,2-triethoxyacetate
Supplier: Apollo Scientific
|
Warning
|
OR939203-1G
OR939203-250MG
OR939203-5G
APOSOR939203-1GEA
58
EUR
APOSOR939203-1G
APOSOR939203-250MG
APOSOR939203-5G
Ethyl 2,2,2-triethoxyacetate
Ethyl 2,2,2-triethoxyacetate
Formula:
C₁₀H₂₀O₅ MW: 220.263 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00449653 CAS Number: 57267-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...