7-Methoxyisochroman-4-one 95%

Supplier: Apollo Scientific

OR942314-1G OR942314-250MG
APOSOR942314-1GEA 288 EUR
APOSOR942314-1G APOSOR942314-250MG
7-Methoxyisochroman-4-one 95%
7-Methoxyisochroman-4-one
Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD09744076
CAS Number: 54212-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR