1-(3-Nitrophenyl)ethanamine 95%
Supplier: Apollo Scientific
OR943499-1G
OR943499-250MG
APOSOR943499-1GEA
137
EUR
APOSOR943499-1G
APOSOR943499-250MG
1-(3-Nitrophenyl)ethanamine 95%
1-(3-Nitrophenyl)ethanamine
Formula:
C₈H₁₀N₂O₂ MW: 166.177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04974122 CAS Number: 90271-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...