2-(4-Chlorophenyl)-1,3,4-oxadiazole 95%

Supplier: Apollo Scientific

OR954505-10G OR954505-1G OR954505-5G
APOSOR954505-10GEA 502 EUR
APOSOR954505-10G APOSOR954505-1G APOSOR954505-5G
2-(4-Chlorophenyl)-1,3,4-oxadiazole 95%
2-(4-Chlorophenyl)-1,3,4-oxadiazole
Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08361748
CAS Number: 23289-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR