5-(3-Chlorophenyl)-3H-1,3,4-oxadiazol-2-one 98%

Supplier: Apollo Scientific

OR954600-10G OR954600-1G OR954600-5G
APOSOR954600-10GEA 574 EUR
APOSOR954600-10G APOSOR954600-1G APOSOR954600-5G
5-(3-Chlorophenyl)-3H-1,3,4-oxadiazol-2-one 98%
5-(3-Chlorophenyl)-3H-1,3,4-oxadiazol-2-one
Formula: C₈H₅ClN₂O₂
MW: 196.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046709
CAS Number: 85929-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR