1-{1-[(4-Fluorophenyl)methyl]-1H-1,2,3-triazol-4-yl}ethan-1-one

Supplier: Apollo Scientific

PC200081-1G PC200081-500MG
APOSPC200081-1GEA 242 EUR
APOSPC200081-1G APOSPC200081-500MG
1-{1-[(4-Fluorophenyl)methyl]-1H-1,2,3-triazol-4-yl}ethan-1-one
1-{1-[(4-Fluorophenyl)methyl]-1H-1,2,3-triazol-4-yl}ethan-1-one
Formula: C₁₁H₁₀FN₃O
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD25368737
CAS Number: 1355359-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR