2-Fluoro-4,6-diiodoaniline

Supplier: Apollo Scientific

PC210036-1G PC210036-500MG
APOSPC210036-1GEA 1070 EUR
APOSPC210036-1G APOSPC210036-500MG
2-Fluoro-4,6-diiodoaniline
2-Fluoro-4,6-diiodoaniline
Formula: C₆H₄FI₂N
MDL Number: MFCD14708138
CAS Number: 1301739-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR