8-Fluoro-3-iodoquinoline 99%
Supplier: Apollo Scientific
|
Warning
|
PC407025-1G
PC407025-250MG
PC407025-5G
APOSPC407025-1GEA
95.7
EUR
APOSPC407025-1G
APOSPC407025-250MG
APOSPC407025-5G
8-Fluoro-3-iodoquinoline 99%
8-Fluoro-3-iodoquinoline
Formula:
C₉H₅FIN MW: 273.046 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07700291 CAS Number: 866782-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...