1-(Difluoromethoxy)-3-fluoro-2-methylbenzene 95%

Supplier: Apollo Scientific

PC421067-10G PC421067-1G PC421067-5G
APOSPC421067-10GEA 1980 EUR
APOSPC421067-10G APOSPC421067-1G APOSPC421067-5G
1-(Difluoromethoxy)-3-fluoro-2-methylbenzene 95%
1-(Difluoromethoxy)-3-fluoro-2-methylbenzene
Formula: C₈H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD14698458
CAS Number: 1214333-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR