1-Chloro-2H-perfluorocyclopentane 95%

Supplier: Apollo Scientific
Warning

PC450580-1G PC450580-5G
APOSPC450580-1GEA 139 EUR
APOSPC450580-1G APOSPC450580-5G
1-Chloro-2H-perfluorocyclopentane 95%
1-Chloro-2H-perfluorocyclopentane
Formula: C₅HClF₈
Storage Temperature: Ambient
MDL Number: MFCD31560919
CAS Number: 2149598-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR