3-Chloro-4-fluoro-5-(trifluoromethyl)benzoic acid 95%

Supplier: Apollo Scientific
Warning

PC48803-1G PC48803-250MG PC48803-5G
APOSPC48803-1GEA 287 EUR
APOSPC48803-1G APOSPC48803-250MG APOSPC48803-5G
3-Chloro-4-fluoro-5-(trifluoromethyl)benzoic acid 95%
3-Chloro-4-fluoro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₃ClF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD13192650
CAS Number: 381229-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR