5-Fluoro-2-vinylbenzeneboronic acid
Supplier: Apollo Scientific
|
Warning
|
PC53466-1G
PC53466-250MG
APOSPC53466-1GEA
638
EUR
APOSPC53466-1G
APOSPC53466-250MG
5-Fluoro-2-vinylbenzeneboronic acid
5-Fluoro-2-vinylbenzeneboronic acid
Formula:
C₈H₈BFO₂ MW: 165.957 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30723298 CAS Number: 1249506-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...