2-(4-Fluorophenyl)-1,3,4-oxadiazole 98%

Supplier: Apollo Scientific

PC909153-10G PC909153-1G PC909153-5G
APOSPC909153-10GEA 502 EUR
APOSPC909153-10G APOSPC909153-1G APOSPC909153-5G
2-(4-Fluorophenyl)-1,3,4-oxadiazole 98%
2-(4-Fluorophenyl)-1,3,4-oxadiazole
Formula: C₈H₅FN₂O
Storage Temperature: Ambient
MDL Number: MFCD11044696
CAS Number: 595567-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR