Perfluoropentadecane 95%

Supplier: Apollo Scientific
Warning

PC9799-5G PC9799-1G
APOSPC9799-5GEA 219 EUR
APOSPC9799-5G APOSPC9799-1G
Perfluoropentadecane 95%
Perfluoropentadecane
Formula: C₁₅F₃₂
MW: 788.109 g/mol
Boiling Pt: 228 °C
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00239439
CAS Number: 2264-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR