1,1,2,2-Tetrachloroethane-[D₂] (99.6% D), NMR SPECTRONORM® for NMR spectroscopy

87094.0010EA 203 EUR
87094.0010 87094.0025
1,1,2,2-Tetrachloroethane-[D₂] (99.6% D), NMR SPECTRONORM® for NMR spectroscopy
1,1,2,2-Tetrachloroethane-[D2]
Formula: Cl₂CDCDCl₂
MW: 169.83 g/mol
Boiling Pt: 146
Melting Pt: -43
MDL Number: MFCD00037672
CAS Number: 33685-54-0

Order Now

Specification Test Results

Purity and Structure by [1H] NMR Conforms to reference
Isotopic enrichment ≥99,6% atom D
Water content <200 ppm
Acidity (pH) >5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR