N-Acetyl-DL-leucine
|
Warning
|
BIA1766-100G
BIA1766-25G
APOSBIA1766-100GEA
16.1
EUR
APOSBIA1766-100G
APOSBIA1766-25G
N-Acetyl-DL-leucine
N-Acetyl-DL-leucine
Formula:
C₈H₁₅NO₃ Melting Pt: 158…160 °C Storage Temperature: Freezer |
MDL Number:
MFCD00026498 CAS Number: 99-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...