7-Aminocephalosporanic acid

Supplier: Apollo Scientific
Danger

BIA4007-10G BIA4007-5G
APOSBIA4007-10GEA 79.9 EUR
APOSBIA4007-10G APOSBIA4007-5G
7-Aminocephalosporanic acid
7-Aminocephalosporanic acid
Formula: C₁₀H₁₂N₂O₅S
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005177
CAS Number: 957-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR