Aflatoxin B1

Danger

BIA4308-10MG BIA4308-5MG BIA4308-1MG
APOSBIA4308-10MGEA 372 EUR
APOSBIA4308-10MG APOSBIA4308-5MG APOSBIA4308-1MG
Aflatoxin B1
Aflatoxin B1

Hepatocarcinogen

Formula: C₁₇H₁₂O₆
Melting Pt: 268…269 °C
Storage Temperature: Refrigerator
CAS Number: 1162-65-8
EINECS: 214-603-3
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR