Aflatoxin B2

Danger

BIA4309-1MG BIA4309-5MG
APOSBIA4309-1MGEA 143 EUR
APOSBIA4309-1MG APOSBIA4309-5MG
Aflatoxin B2
Aflatoxin B2

Hepatocarcinogen

Formula: C₁₇H₁₄O₆
Melting Pt: 286…289 °C
Storage Temperature: Refrigerator
CAS Number: 7220-81-7
EINECS: 230-618-8
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR