Aflatoxin G1

Danger

BIA4310-1MG BIA4310-5MG
APOSBIA4310-1MGEA 147 EUR
APOSBIA4310-1MG APOSBIA4310-5MG
Aflatoxin G1
Aflatoxin G1

Hepatocarcinogen

Formula: C₁₇H₁₂O₇
Melting Pt: 244…246 °C
Storage Temperature: Refrigerator
CAS Number: 1165-39-5
EINECS: 214-615-9
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR