Aflatoxin G2

Danger

BIA4311-1MG BIA4311-5MG
APOSBIA4311-1MGEA 328 EUR
APOSBIA4311-1MG APOSBIA4311-5MG
Aflatoxin G2
Aflatoxin G2

Hepatocarcinogen

Formula: C₁₇H₁₄O₇
Storage Temperature: Refrigerator
CAS Number: 7241-98-7
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR