Dextran sulphate sodium salt, M.W. 15000

Warning

BID1343-100G BID1343-25G BID1343-50G
APOSBID1343-100GEA 165 EUR
APOSBID1343-100G APOSBID1343-25G APOSBID1343-50G
Dextran sulphate sodium salt, M.W. 15000
Dextran sulphate sodium salt
Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR