Quizartinib 99%

Warning

BIFK0001-1MG BIFK0001-5MG
APOSBIFK0001-1MGEA 14.7 EUR
APOSBIFK0001-1MG APOSBIFK0001-5MG
Quizartinib 99%
Quizartinib

  • Free Base, >99%

Formula: C₂₉H₃₂N₆O₄S
Storage Temperature: Freezer
MDL Number: MFCD18074524
CAS Number: 950769-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR