Avasimibe
BISN0114-5MG
BISN0114-10MG
BISN0114-50MG
APOSBISN0114-5MGEA
109
EUR
APOSBISN0114-5MG
APOSBISN0114-10MG
APOSBISN0114-50MG
Avasimibe
Avasimibe
Avasimibe is an ACAT inhibitor with IC50 of 33 M it also inhibits human P450 isoenzymes CYP2C9 CYP1A2 and CYP2C19 with IC50 of 29139 and 265 M respectively.
Formula:
C₂₉H₄₃NO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00934956 CAS Number: 166518-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...