T-2 Toxin

Danger

BIT1041-1MG BIT1041-5MG
APOSBIT1041-1MGEA 87.3 EUR
APOSBIT1041-1MG APOSBIT1041-5MG
T-2 Toxin
T-2 Toxin
Melting Pt: 151…152 °C
Storage Temperature: Freezer
CAS Number: 21259-20-1
EINECS: 244-297-7
UN: 3462
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR