4-Ethynylbenzaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR01516-1G
OR01516-25G
OR01516-5G
APOSOR01516-1GEA
21.9
EUR
APOSOR01516-1G
APOSOR01516-25G
APOSOR01516-5G
4-Ethynylbenzaldehyde 97%
4-Ethynylbenzaldehyde
Formula:
C₉H₆O MW: 130.146 g/mol Melting Pt: 89…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664348 CAS Number: 63697-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...