5-Pyrimidinecarbonitrile

Supplier: Apollo Scientific
Warning

OR11333-5G OR11333-1G
APOSOR11333-5GEA 171 EUR
APOSOR11333-5G APOSOR11333-1G
5-Pyrimidinecarbonitrile
5-Pyrimidinecarbonitrile
Formula: C₅H₃N₃
Storage Temperature: Ambient
MDL Number: MFCD00235052
CAS Number: 40805-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR