(4-Chlorophenylsulphonyl)urea 98%
Supplier: Apollo Scientific
|
Warning
|
OR1360-100G
OR1360-25G
OR1360-5G
APOSOR1360-100GEA
621
EUR
APOSOR1360-100G
APOSOR1360-25G
APOSOR1360-5G
(4-Chlorophenylsulphonyl)urea 98%
(4-Chlorophenylsulphonyl)urea
Formula:
C₇H₇ClN₂O₃S Melting Pt: 169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00518656 CAS Number: 22663-37-2 EINECS: 245-149-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...