1-(2,3-Dihydro-1-benzofuran-5-ylmethyl)-4-piperidinecarboxylic acid

Supplier: Apollo Scientific

OR15080-1G OR15080-5G
APOSOR15080-1GEA 538 EUR
APOSOR15080-1G APOSOR15080-5G
1-(2,3-Dihydro-1-benzofuran-5-ylmethyl)-4-piperidinecarboxylic acid
1-(2,3-Dihydro-1-benzofuran-5-ylmethyl)-4-piperidinecarboxylic acid
Formula: C₁₅H₁₉NO₃
MW: 261.32 g/mol
Melting Pt: 278…281 °C
Storage Temperature: Ambient
MDL Number: MFCD03645498
CAS Number: 887440-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR