2-Aminobenzanilide 97%

Supplier: Apollo Scientific
Warning

OR1564-25G OR1564-5G OR1564-1G
APOSOR1564-25GEA 606 EUR
APOSOR1564-25G APOSOR1564-5G APOSOR1564-1G
2-Aminobenzanilide 97%
2-Aminobenzanilide
Formula: C₁₃H₁₂N₂O
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00034792
CAS Number: 4424-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR