4-Phenylbenzophenone 99%
Supplier: Apollo Scientific
|
Warning
|
OR18324-100G
OR18324-500G
APOSOR18324-100GEA
36.4
EUR
APOSOR18324-100G
APOSOR18324-500G
4-Phenylbenzophenone 99%
4-Phenylbenzophenone
Formula:
C₁₉H₁₄O Boiling Pt: 419…420 °C Melting Pt: 98…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003079 CAS Number: 2128-93-0 EINECS: 218-345-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...