2-Aminothiophenol 98%

Supplier: Apollo Scientific
Danger

OR1999-100G OR1999-500G
APOSOR1999-100GEA 128 EUR
APOSOR1999-100G APOSOR1999-500G
2-Aminothiophenol 98%
2-Aminothiophenol
Formula: C₆H₇NS
Boiling Pt: 234 °C
Melting Pt: 16…20 °C
Density: 1,17 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR