5-Bromosalicylaldehyde 98%

Supplier: Apollo Scientific
Warning

OR21356-100G OR21356-1KG OR21356-500G
APOSOR21356-100GEA 21.9 EUR
APOSOR21356-100G APOSOR21356-1KG APOSOR21356-500G
5-Bromosalicylaldehyde 98%
5-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
Melting Pt: 102…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR