Levoglucosenone
Supplier: Apollo Scientific
|
Warning
|
OR22203-1G
OR22203-250MG
OR22203-5G
APOSOR22203-1GEA
400
EUR
APOSOR22203-1G
APOSOR22203-250MG
APOSOR22203-5G
Levoglucosenone
Levoglucosenone
Formula:
C₆H₆O₃ MW: 126.11 g/mol Boiling Pt: 115 °C/10mmHg Storage Temperature: Freezer |
MDL Number:
MFCD00191664 CAS Number: 37112-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...