3-Chlorobenzylideneacetone
Supplier: Apollo Scientific
|
Warning
|
OR24012-25G
OR24012-5G
APOSOR24012-25GEA
0
EUR
APOSOR24012-25G
APOSOR24012-5G
3-Chlorobenzylideneacetone
3-Chlorobenzylideneacetone
Formula:
C₁₀H₉ClO Boiling Pt: 123…125 °C/1mmHg Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052851 CAS Number: 20766-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...