3-Oxetanone 98%
Supplier: Apollo Scientific
|
Warning
|
OR301011-25G
OR301011-5G
APOSOR301011-25GEA
45.1
EUR
APOSOR301011-25G
APOSOR301011-5G
3-Oxetanone 98%
3-Oxetanone
Formula:
C₃H₄O₂ Boiling Pt: 140 °C Density: 1,124 g/cm³ (25 °C) Flash Pt: 26 °C Storage Temperature: Freezer |
MDL Number:
MFCD09263255 CAS Number: 6704-31-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...