Isopropylsulphamoyl chloride

Supplier: Apollo Scientific
Danger

OR346199-1G OR346199-5G
APOSOR346199-1GEA 306 EUR
APOSOR346199-1G APOSOR346199-5G
Isopropylsulphamoyl chloride
Isopropylsulphamoyl chloride
Formula: C₃H₈ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD09753748
CAS Number: 26118-67-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR