N-Methylallylamine

Supplier: Apollo Scientific
Danger

OR3619-25G OR3619-5G
APOSOR3619-25GEA 26.1 EUR
APOSOR3619-25G APOSOR3619-5G
N-Methylallylamine
N-Methylallylamine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 61…66 °C
Density: 0.748 g/cm³ (25 °C)
Flash Pt: –8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008641
CAS Number: 627-37-2
EINECS: 210-996-0
UN: 3286
ADR: 3+6.1+8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR