N,N-Diispropylbenzamide 98%
Supplier: Apollo Scientific
|
Warning
|
OR451332-25G
OR451332-5G
APOSOR451332-25GEA
743
EUR
APOSOR451332-25G
APOSOR451332-5G
N,N-Diispropylbenzamide 98%
N,N-Diispropylbenzamide
Formula:
C₁₃H₁₉NO Melting Pt: 69…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026370 CAS Number: 20383-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...