4-Benzyloxy-3-methylphenylboronic acid 96%

Supplier: Apollo Scientific
Warning

OR5541-1G OR5541-5G
APOSOR5541-1GEA 0 EUR
APOSOR5541-1G APOSOR5541-5G
4-Benzyloxy-3-methylphenylboronic acid 96%
4-Benzyloxy-3-methylphenylboronic acid
Formula: C₁₄H₁₅BO₃
Melting Pt: 102…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409944
CAS Number: 338454-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR