3-Bromo-N,N-diethylbenzenesulphonamide 98%
Supplier: Apollo Scientific
|
Warning
|
OR5725-10G
OR5725-1G
OR5725-5G
APOSOR5725-10GEA
716
EUR
APOSOR5725-10G
APOSOR5725-1G
APOSOR5725-5G
3-Bromo-N,N-diethylbenzenesulphonamide 98%
3-Bromo-N,N-diethylbenzenesulphonamide
Formula:
C₁₀H₁₄BrNO₂S Melting Pt: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363825 CAS Number: 871269-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...