4-Hydroxyphthalic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR59816-25G
OR59816-5G
OR59816-100G
APOSOR59816-25GEA
50.9
EUR
APOSOR59816-25G
APOSOR59816-5G
APOSOR59816-100G
4-Hydroxyphthalic acid 98%
4-Hydroxyphthalic acid
Formula:
C₈H₆O₅ Melting Pt: 196…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013984 CAS Number: 610-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...