Pivalonitrile 98%

Supplier: Apollo Scientific
Danger

OR6535-100G OR6535-25G OR6535-5G OR6535-1G
APOSOR6535-100GEA 205 EUR
APOSOR6535-100G APOSOR6535-25G APOSOR6535-5G APOSOR6535-1G
Pivalonitrile 98%
Pivalonitrile
Formula: C₅H₉N
MW: 83.13 g/mol
Boiling Pt: 105…106 °C
Melting Pt: 15…16 °C
Density: 0.752 g/cm³ (25 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001847
CAS Number: 630-18-2
EINECS: 211-133-0
UN: 3273
ADR: 3+6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR