4-(2-Phenylethylcarbamoyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

OR7005-1G OR7005-5G
APOSOR7005-1GEA 49.3 EUR
APOSOR7005-1G APOSOR7005-5G
4-(2-Phenylethylcarbamoyl)phenylboronic acid 98%
4-(2-Phenylethylcarbamoyl)phenylboronic acid
Formula: C₁₅H₁₆BNO₃
Melting Pt: 195…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09027209
CAS Number: 330793-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR