2-Methoxycyclohexanone 95%
Supplier: Apollo Scientific
OR920090-1G
OR920090-25G
OR920090-5G
APOSOR920090-1GEA
109
EUR
APOSOR920090-1G
APOSOR920090-25G
APOSOR920090-5G
2-Methoxycyclohexanone 95%
2-Methoxycyclohexanone
Formula:
C₇H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00009610 CAS Number: 7429-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...