3-Amino-5-chlorothiophene hydrochloride 95%

Supplier: Apollo Scientific

OR921584-1G OR921584-250MG
APOSOR921584-1GEA 719 EUR
APOSOR921584-1G APOSOR921584-250MG
3-Amino-5-chlorothiophene hydrochloride 95%
3-Amino-5-chlorothiophene hydrochloride
Formula: C₄H₅Cl₂NS
Storage Temperature: Ambient
MDL Number: MFCD20926161
CAS Number: 1408076-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR