Benzoylnitromethane 95%
Supplier: Apollo Scientific
|
Warning
|
OR921784-1G
OR921784-25G
OR921784-5G
APOSOR921784-1GEA
109
EUR
APOSOR921784-1G
APOSOR921784-25G
APOSOR921784-5G
Benzoylnitromethane 95%
Benzoylnitromethane
Formula:
C₈H₇NO₃ Melting Pt: 105…107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010218 CAS Number: 614-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...