6-Nitrochromone 95%

Supplier: Apollo Scientific

OR925526-25G OR925526-5G
APOSOR925526-25GEA 526 EUR
APOSOR925526-25G APOSOR925526-5G
6-Nitrochromone 95%
6-Nitrochromone
Formula: C₉H₅NO₄
MDL Number: MFCD02954226
CAS Number: 51484-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR